Orders LinkContactUsLinkComponent
SEC standards, AdvanceBio
SEC standards, AdvanceBio
RNA resolution standard, oligos at 14, 17, 20, and 21 mer, 1
Codice catalogo HEWL5190-9028
Nome del fornitore:   AGILENT HEWLETT PACKARD
SEC standards, AdvanceBio
Codice catalogo HEWL5190-9028
Nome del fornitore:  AGILENT HEWLETT PACKARD
Numero CAS:  
SEC standards, AdvanceBio
SEC standards, AdvanceBio
RNA resolution standard, oligos at 14, 17, 20, and 21 mer, 1
Numero CAS:  
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
AdvanceBio SEC Protein Standards are uniquely designed for AdvanceBio SEC columns. The standards are manufactured from a select group of proteins and peptides optimized to deliver accurate, precise quantitation for mAb aggregation and protein analysis.
  • Wide range of molecular weight proteins
  • Optimize for use with AdvanceBio SEC columns for maximum peak to peak resolution
  • Manufactured and shipped in moisture and light resistant packaging to ensure stability of standard
This new SEC technology is designed and engineered by Agilent to improve lab productivity by providing robust, reliable methods that eliminate sample reanalysis. Consistent results are achieved from column to column, batch to batch, and lab to lab, ensuring that methods can be transferred across departments and locations to put an end to uncertainty.
Specifiche
Specifiche
  • Conf:
    1
  • Descrizione:
    RNA resolution standard, oligos at 14, 17, 20, and 21 mer