Earplugs dispenser, Leight® Source 500
Earplugs without cord, refill for dispenser Leight® Source 500, 37 dB, arancione
Codice catalogo DALL3301165
Nome del fornitore: HONEYWELL SAFETY
Earplugs dispenser, Leight® Source 500
Codice catalogo DALL3301165
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
The Leight Source 500 dispenser provides a highly visible permanent hearing protection source.
- Heavy-duty anodized aluminum allowing constant use
- Wall mounting for a user-friendly access
- Crank handle to dispense earplugs
Suitable for large factories and process industries with Max®, Max Lite®, Laser Lite®, Multi Max®, Matrix™, FirmFit™, Quiet® earplugs.
Specifiche
- Colore:arancione
- Descrizione:Earplugs without cord, refill for dispenser Leight® Source 500, 37 dB
- Seq:04
- Cat. No.:DALL3301165
- Tipo di dispenser:Montabile a parete
- PPE type:Tappi auricolari