Orders LinkContactUsLinkComponent
Drum racks
Drum racks
Type 120, with GRP sump pallet (65 l) and grating support for small containers, heights of storage levels from ground 600/1120 mm, 21 kg, 450×820×1310 mm
Codice catalogo CEMG7639
Nome del fornitore:   CEMO
Drum racks
Codice catalogo CEMG7639
Nome del fornitore:  CEMO
Drum racks
Drum racks
Type 120, with GRP sump pallet (65 l) and grating support for small containers, heights of storage levels from ground 600/1120 mm, 21 kg, 450×820×1310 mm
Price
SFr. 663.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Drum racks for approved and space saving storage of barrels and other containers.
  • All drum racks are completely hotdip galvanised and have a corrosion free GRP sump pallet for long-term safe use
  • With general construction inspection approval no. Z-40.12-227 granted by DIBt-Berlin
  • Universally applicable for waste oil, lyes and acids
Specifiche
  • Peso:
    21 kg
  • Descrizione:
    Type 120, with GRP sump pallet (65 l) and grating support for small containers, heights of storage levels from ground 600/1120 mm
  • Seq:
    03
  • L×P×A:
    450×820×1310 mm
Accessories
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mm
    Can holder, galvanised, for drum racks type 120/360/540, with surface 230×110 mm
    CEMG5917
    2 kg
    Log in per disponibilità
    1 * 1 ST - 49.20 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
    Rotating roller support for drum racks type 400/800 (for 200 l barrel (pair), heights of storage levels 660 mm from ground
    CEMG7669
    6 kg
    Log in per disponibilità
    1 * 1 ST - 282.00 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums
    Drum supports for drum racks for type 360/540, 3 pieces for one storage level for optimum emptying of the 60 l drums
    CEMG8254
    1 kg
    Log in per disponibilità
    1 * 3 ST - 61.60 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from ground
    Steel grating for drum racks, as 3rd (for type 360) or 4th storage level (for type 540), 1300×620×30 mm, 210 mm storage height from ground
    CEMG6817
    18 kg
    Log in per disponibilità
    1 * 1 ST - 153.00 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
    Can holder, galvanised, for drum racks type 400/800, with bearing surface 185×110 mm
    CEMG7727
    2 kg
    Log in per disponibilità
    1 * 1 ST - 56.20 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    Disponibilità
    Price
  • Drum supports, 3 pieces per storage level, for drum racks type 360/540
    Drum supports, 3 pieces per storage level, for drum racks type 360/540
    CEMG7800
    1 kg
    Log in per disponibilità
    1 * 1 ST - 57.90 SFr.
Informazioni sul prodotto
  • Codice catalogo
    Peso
    L×P×A
    Disponibilità
    Price
  • Sump pallet, PE, 40 l, without grating
    Sump pallet, PE, 40 l, without grating
    CEMG7871
    5 kg
    1300×620×80 mm
    Log in per disponibilità
    1 * 1 ST - 196.00 SFr.