Microplate handling system, StakMax®
20-plate stacker magazines (pair)
Codice catalogo MLDV0200-6016
Nome del fornitore: Molecular Devices
Microplate handling system, StakMax®
Codice catalogo MLDV0200-6016
Nome del fornitore: Molecular Devices
Vai a
Product Details & Documents
The StakMax® microplate handling system is a compact benchtop microplate stacker that works with microplate readers and microplate washers to give walk-away automation for up to 50 microplates.
- Integrates with microplate readers and AquaMax® washers for improved process flow and higher throughput
- Flexible operation using powerful SoftMax® Pro software
- Barcode reader can be added to enable barcode identification of plates
- Minimises plate movement time, while handling the microplates gently, eliminating liquid spillage
Specifiche
- Descrizione:20-plate stacker magazines (pair)
- Seq:05