Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Quick-DNA™ Plus Kits

Quick-DNA™ Miniprep Plus kit, 200 Preps, 1 KIT
Codice catalogo ZYMOD4069
Marketsource
Nome del fornitore:   Zymo Research
Quick-DNA™ Plus Kits
Codice catalogo ZYMOD4069
Nome del fornitore:  Zymo Research
Marketsource
Numero CAS:  
Quick-DNA™ Plus Kits
Quick-DNA™ Plus Kits

Quick-DNA™ Miniprep Plus kit, 200 Preps, 1 KIT

Numero CAS:  
Price
SFr. 746.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents

DNA from any sample (cells, tissues, biological fluids and more).

  • Quick and easy: Simple 20 minute procedure
  • Highest yield: Recover 3× more DNA
  • Ultra-pure: Ready for qPCR, next-generation sequencing, arrays and more

The Quick-DNA™ plus kits are the easiest method for high yield total DNA extraction (e.g., genomic, mitochondrial, viral) from any biological fluid, cell culture, or solid tissue sample. Innovative reagents and zymo-spin technology allow for ultra-pure and concentrated genomic DNA >50 kb to be eluted in as little as 10 µl. Zymo-Spin™ columns ensure no buffer retention. Purified DNA is RNA-free bypassing the need for RNase A treatment and ensuring accurate quantification for applications like library preparations. Isolated DNA is suitable for immediate use in sensitive downstream applications including qPCR, DNA-seq, arrays and methylation analysis.

Specifiche

Specifiche

  • Conf:
    1 KIT
  • Descrizione:
    Quick-DNA™ Miniprep Plus kit
  • Volume:
    200 Preps
  • Cat. No.:
    ZYMOD4069
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Target molecule:
    DNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Blood|Biofluids (serum, plasma, saliva)|Cultured cells|Human/animal cells and tissues
  • Resa:
    The DNA binding capacity of the column is 25 µg. Typically, mammalian tissues yield: 1-3 µg DNA per mg skeletal, heart, lung, and brain tissues and 3-5 µg DNA per mg liver and kidney. Human whole blood will yield 3-7 µg DNA per 100 µl blood sampled.
  • Descrizione:
    Quick-DNA™ Miniprep Plus kit
  • Capacità campioni:
    25 µg
  • Dimensione del campione:
    ≤5×10⁶ Cells or 25 mg Tissue

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..