Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Detectable spoons, SteriPlast®

Detectable spoons, SteriPlast®

170 mm, 10 ml
Catalog # 231-0576
Supplier:   BURKLE
Detectable spoons, SteriPlast®
Catalog # 231-0576
Supplier:  BURKLE
Detectable spoons, SteriPlast®
Detectable spoons, SteriPlast®

170 mm, 10 ml

Price
SFr. 90.90
Log in for availability
1 * 100 ST
Product Details & Documents

These detectable SteriPlast® spoons, made of PS, are produced in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The detectable spoons are available in two sizes, 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).

  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations

These spoons are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.

Specifications
  • Capacità:
    10 ml
  • Lunghezza:
    170 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.