Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Detectable spoons, SteriPlast®

127 mm, 2,5 ml
Codice catalogo 231-0575
Nome del fornitore:   BURKLE
Detectable spoons, SteriPlast®
Codice catalogo 231-0575
Nome del fornitore:  BURKLE
Detectable spoons, SteriPlast®
Detectable spoons, SteriPlast®

127 mm, 2,5 ml

Price
SFr. 82.00
Log in per disponibilità
1 * 100 ST
Product Details & Documents

These detectable SteriPlast® spoons, made of PS, are produced in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The detectable spoons are available in two sizes, 2,5 ml (similar to a teaspoon) and 10 ml (similar to a tablespoon).

  • Can be detected by colour scanners, metal detectors or X-rays
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations

These spoons are not only coloured blue but also have a special additive in the material that makes them visible to metal detectors or X rays. These products can therefore be rejected with standard systems for checking for foreign objects, even though they are made of plastic.

Specifiche
  • Capacità:
    2,5 ml
  • Lunghezza:
    127 mm
  • Seq:
    01