Accessories for microcarriers
Aseptic transfer cap, PP, orange, disposable ,48 mm, male luer lock, for 500 g microcarrier bottle
734-2921
: Corning
These polypropylene aseptic transfer screw caps prevents adventitious contamination in for microcarriers bottles during bioreactor loading.
- Chemically resistant and heat sealable flexible tubing
- Tubing length with connector 30" (76,2 cm)
- Disposable and sterile (10⁻⁶ SAL)
: These caps are individually double bagged.
: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
- Descrizione:Aseptic transfer cap, PP, orange, disposable ,48 mm, male luer lock, for 500 g microcarrier bottle
- Cat. No.:734-2921
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:RECs, ISO 14001 - Kennebunk