Accessories for CryoPlus Sample Storage Systems
Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
Codice catalogo THBR4000042
Nome del fornitore: Thermo Fisher Scientific
Accessories for CryoPlus Sample Storage Systems
Codice catalogo THBR4000042
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
Specifiche
- Descrizione:Rack includes cardboard boxes and cardboard dividers for 1,5/2 ml vials, 660×140×143 mm, for CryoPlus 1, 2, 4
- Seq:15
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.