Orders LinkContactUsLinkComponent
Novec™ 7100 engineered fluid | Avantor
Novec™ 7100 engineered fluid
Poly bottle, 1 L
Codice catalogo SHH0002-1L
Nome del fornitore:   SIGMA ALDRICH MICROSCOPY
Novec™ 7100 engineered fluid
Codice catalogo SHH0002-1L
Nome del fornitore:  SIGMA ALDRICH MICROSCOPY
Numero CAS:  163702-08-7;163702-07-6
Novec™ 7100 engineered fluid
Novec™ 7100 engineered fluid
Poly bottle, 1 L
Numero CAS:  163702-08-7;163702-07-6
Price
SFr. 328.00
Log in per disponibilità
1 * 1 L
Product Details & Documents
Novec™ 7100 fluid consists of two inseparable isomers ((CF₃)₂CFCF₂OCH₃) and CF₃CF₂CF₂CF₂OCH₃) with essentially identical properties. Ideal for vapor degreasing applications as a neat (pure), azeotropic component and co-solvent parts cleaner.
  • Non-flammable and non-conductive
  • Compatible with plastics
  • Low toxicity and surface tension
  • Low global warming potential (GWP) and zero ozone depletion potential
  • Histology Bath Fluid
Used as a heat transfer fluid, cleaning and rinsing agent, spray contact cleaner, cryofreezing solution, lubricant carrier, dielectric test media and replacement for CFC, HCFC, HFC and PFC.
Novec is a trademark of 3M Company
  • Numero CAS:163702-08-7;163702-07-6
  • Formula:C₅H₃OF₉
  • Numero MDL:MFCD28505595
  • Molecular Weight:250.06 g/mol
Dokumente
Specifiche
Specifiche
  • Tipo di conf.:
    Poly bottle
  • Conf:
    1 L
Specifiche