Flasks, round bottom, Quickfit® ground joints
2000 ml, 34/35 NS, 228 mm
Codice catalogo BIBBFR2L/5S
Nome del fornitore: SciLabware
Flasks, round bottom, Quickfit® ground joints
Codice catalogo BIBBFR2L/5S
Nome del fornitore: SciLabware
Vai a
Product Details & Documents
PYREX®, borosilicate glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
: ISO 1773, ISO 4797.
Specifiche
- Cono femmina:34/35 NS
- Capacità:2000 ml
- Altezza:228 mm
- Seq:40



