Orders LinkContactUsLinkComponent
Fractionating columns, Vigreux, Quickfit®
600 mm, 24/29 NS, 24/29 NS
Codice catalogo BIBBFC3/23
Nome del fornitore:   SciLabware
Fractionating columns, Vigreux, Quickfit®
Codice catalogo BIBBFC3/23
Nome del fornitore:  SciLabware
Fractionating columns, Vigreux, Quickfit®
Fractionating columns, Vigreux, Quickfit®
600 mm, 24/29 NS, 24/29 NS
Price
SFr. 1’920.00
Log in per disponibilità
1 * 5 ST
Product Details & Documents
PYREX®, borosilicate glass
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
No glass jacket. With combination of horizontal and inclined indentations. Horizontal indentations ensure intimate vapour and piquid contatc. Inclined indentations redistribute liquid from walls to centre of column.
Specifiche
  • Dimensione cono:
    24/29 NS
  • Cono femmina:
    24/29 NS
  • Lunghezza utile:
    600 mm
  • Seq:
    03