Cylinders, borosilicate glass, with hexagonal base
5 ml
Codice catalogo SCOT213910776
Nome del fornitore: DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Codice catalogo SCOT213910776
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
- Without graduation
- Hexagonal base
- Base with three knobs which increase its stability
Certifications: DIN EN ISO 4788.
Specifiche
- Capacità:5 ml
- Seq:01