Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
ALLN

ALLN

Codice catalogo ABCAAB141445-5
Nome del fornitore:   Abcam
ALLN
Codice catalogo ABCAAB141445-5
Nome del fornitore:  Abcam
Numero CAS:  
Numero CAS:  
Price
SFr. 180.00
Log in per disponibilità
1 * 5 mg
Product Details & Documents

MW 383.5 Da, Purity >95%. ALLN, cysteine proteinase inhibitor. Achieve your results faster with highly validated, pure and trusted compounds.

  • Constituents: 100% ALLN

Attenzione: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifiche

Specifiche

  • Peso molecolare:
    383.5 DA
  • Cat. No.:
    ABCAAB141445-5
  • Condizioni di conservazione:
    Store at -20 °C.
  • Protein/peptide name:
    ALLN
  • Purezza:
    >95%
  • Temperatura di spedizione:
    Ambient - Can Ship with Ice
  • CAS No.:
    5 mg

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..