ESD insole, ESD-PRO
Insoles, ESD-Pro, full-length, 40, nero
Codice catalogo 114-7005
Nome del fornitore: ELTEN
Vai a
Product Details & Documents
Antibacterial and anatomically shaped ESD insole.
- Open cell structure of PU foam guarantees high moisture absorption
- Activated carbon absorber reduces odour
- Skin friendly materials improve the foot climate
For use with L10, Biomex (Renzo Biomex), Trainers, Sportics, Classic-Line, Men at Work, Safety-Grip, Business (Ronny), White. Suitable for ESD safety shoes (electrical resistance of 105 to 107 Ohm).
Colour: Black
Specifiche
- Taglia scarpe:40
- Colore:nero
- Descrizione:Insoles, ESD-Pro, full-length