Combustion boats
120×12×16 mm
Catalog # TSLA75147
Supplier: TECHNICAL QUARTZ SOLUTIONS
Jump to:
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifications
- Seq:10
- L×P×A:120×12×16 mm