Combustion boats
50×12×5 mm
Catalog # TSLA75142
Supplier: TECHNICAL QUARTZ SOLUTIONS
Jump to:
Product Details & Documents
Combustion boats, quartz, glazed, with or without handle.
- Very good chemical resistance
- Very good temperature resistance
Specifications
- Seq:05
- L×P×A:50×12×5 mm