Orders LinkContactUsLinkComponent
Spill kits, ADR
Large ADR chemical spill kit, 57 litres, includes 35 pads (41×51 mm), 1 socs (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (230×230 mm), 2 yellow disposal
bags and 1 pair of gloves
  121-0193
 :   Brady
Spill kits, ADR
  121-0193
 :  Brady
Spill kits, ADR
Spill kits, ADR
Large ADR chemical spill kit, 57 litres, includes 35 pads (41×51 mm), 1 socs (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (230×230 mm), 2 yellow disposal
bags and 1 pair of gloves
SFr. 182.00
1 * 1 KIT
ADR spill kits are eco-friendly and portable and come in a durable see-through bag with shoulder strap. The small ADR chemical kit is designed for small spills of chemical and hazardous liquid. The large ADR maintenance kit is designed for small spills of industrial oil, water, solvents and coolants. The Re-Form ADR kit S is designed for small spills of industrial liquids except for highly aggressive acids and caustics.
  • Quick and easy access to the sorbents
  • Durable see-through bag makes it easy to check the spill response supplies
  • Descrizione:
    Large ADR chemical spill kit, 57 litres, includes 35 pads (41×51 mm), 1 socs (76×1220 mm), 1 soc (76×2440 mm), 1 pillow (430×480 mm), 4 pillows (230×230 mm), 2 yellow disposal
    bags and 1 pair of gloves
  • Seq:
    02