Bench mounted gas taps
Helium, EPDM seal
KOTT733-00061
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
- Descrizione:Helium, EPDM seal
- Seq:05
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.