Bench mounted gas taps
Argon, EPDM seal
Codice catalogo KOTT733-00060
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
Specifiche
- Descrizione:Argon, EPDM seal
- Seq:03