Orders LinkContactUsLinkComponent
FastPrep® Empty Tubes and Caps
FastPrep® Empty Tubes and Caps
Screw cap with O-ring, orange
Catalog # ICNA115063005
Supplier:   MP Biomedicals
FastPrep® Empty Tubes and Caps
Catalog # ICNA115063005
Supplier:  MP Biomedicals
FastPrep® Empty Tubes and Caps
FastPrep® Empty Tubes and Caps
Screw cap with O-ring, orange
Price
448.00 SFr.
Log in for availability
1 * 1.000 ST
Product Details & Documents
These natural, non-skirted empty Fasrprep® tubes made of PP are used in Fastprep system, to hold sample for homogenisation.
Specifications
  • Descrizione:
    Screw cap with O-ring, orange
  • Seq:
    02