Orders LinkContactUsLinkComponent
Glicina ≥99.5%
Glicina ≥99.5%
250 g
  ICNA04808873
Glicina ≥99.5%
  ICNA04808873
 :  56-40-6
Glicina ≥99.5%
Glicina ≥99.5%
250 g
 :  56-40-6
SFr. 114.00
1 * 250 g
Electrophoresis Grade Heavy metals (as Pb): 20 ppm Used for preparation of Tris-glycine buffers.
 :  Aminoacetic acid, Glycocoll, H-Gly-OH
  •  :56-40-6
  •  :NH₂CH₂COOH
  •  :MFCD00008131
  •  :75.07 g/mol
  •  :Ambiente
Documents
  • Conf:
    250 g