1,3-Propansultone, semi-solid
Glass bottle for solids, 50 g
Codice catalogo ICNA0215639150
Nome del fornitore: MP Biomedicals
1,3-Propansultone, semi-solid
Codice catalogo ICNA0215639150
Nome del fornitore: MP Biomedicals
Numero CAS: 1120-71-4
Vai a
Product Details & Documents
1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.
1,3-Propane Sultone was used as quaternisation agent to transit the PDMAEMA into poly(zwitterions) through surface reaction by treating membranes under a mild condition.
Soluble in cold water, hot water; readily in ketones, esters and aromatic hydrocarbons; insoluble in aliphatic hydrocarbons.
Sinonimi: 1,2-Oxathiolane 2,2-dioxide
- Numero CAS:1120-71-4
- Formula:C₃H₆O₃S
- Numero MDL:MFCD00005355
- Molecular Weight:122.14 g/mol
- Punto di ebollizione:298 °C (1013 hPa)
- Punto di fusione:30…33 °C (lett.)
- Densità:1,392 g/ml
- temperatura di conservazione:Refrigerata
- EINECS:214-317-9
- UN:2811
- ADR:6.1,III
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:50 g



