VWR®, Stoppers, PE
naturale, 13 mm
Codice catalogo 212-0664
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
- Choice of colours
Specifiche
- Ø superiore:13 mm
- Colore:naturale
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



