Columns, PD MiniTrap™ G-10
Product Details & Documents
PD MiniTrap G-10 columns are prepacked, single-use gravity columns for buffer exchange and cleanup of biological samples, for example peptides, small proteins, and oligosaccharides.
- Easy cleanup of peptides, small proteins, or carbohydrates larger than Mr 700 before downstream analysis such as liquid chromatography, electrophoresis, and MS
- Efficient removal of contaminants such as salts, dyes, and radioactive labels
- High desalting capacity
- Preparation of samples in the volume range of 100 to 300 μl
PD MiniTrap G-10 gravity columns give fast and simple desalting and buffer exchange without any need for a purification system. PD MiniTrap G-10 columns are a member of the Trap platform, which addresses the need for flexible, small-scale preparation of proteins and peptides before analysis.
For larger scale please use PD MidiTrap G-10 columns.
: Instructions are supplied with the columns.
Specifiche
- Capacità:2,1 ml
- Descrizione:PD MiniTrap™ G-10
- Colonna:PP
- Dimensione delle sfere:9,7×28 mm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



