Tute, Secutex® pro 5/6
bianco, XXL
Codice catalogo ZVGA30445-90
Nome del fornitore: Zellstoff-Vertriebs-GmbH & Co. KG
Tute, Secutex® pro 5/6
Codice catalogo ZVGA30445-90
Nome del fornitore: Zellstoff-Vertriebs-GmbH & Co. KG
Vai a
Product Details & Documents
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
- Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
- Permeable to air and water vapour
- Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
Certifications: CE 0624, Codice III, Tipo 5 + 6.
Specifiche
- Taglia dell'abbigliamento:XXL
- Colore:bianco
- Seq:04
- Cat. No.:ZVGA30445-90
- Antistatico:+
- Applicazioni per abiti:Chemical protection|Laboratory|Industrial