Orders LinkContactUsLinkComponent
VWR® Prague, Safety Spectacles
Anti-UV, anti-scratch, anti-fog, blu, Clear PC
  111-1846
VWR® Prague, Safety Spectacles
  111-1846
VWR® Prague, Safety Spectacles
VWR® Prague, Safety Spectacles
Anti-UV, anti-scratch, anti-fog, blu, Clear PC
20.20 SFr.
1 * 1 ST
These economical, compact cover glasses (OTG) fit over most prescription eyewear.
  • Pantoscopic adjustment allows lens angle modification
  • Adjustable temple length
 : Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)
  • Tipo:
    Anti-UV, anti-scratch, anti-fog
  • Colore:
    blu
  • Materiale lente:
    Clear PC
Accessories
  •  
    Descrizione
  • 111-2380
    Spectacle loop cord, black
    1 * 10 ST - 7.00 SFr.
  •  
    Descrizione
    L×L
    Confezione
  • 111-5004
    Lens cleaning tissues
    100×150 mm
    D: 25×25
    1 * 625 ST - 132.00 SFr.
  •  
    Descrizione
  • 111-2382
    Spectacle cord
    1 * 10 ST - 14.00 SFr.
  •  
    Descrizione
  • 111-2383
    Spectacle cloth, black
    1 * 10 ST - 9.20 SFr.
  •  
    Descrizione
  • 111-2381
    Spectacle loop cord with stopper, black
    1 * 10 ST - 16.20 SFr.
  •  
    Descrizione
  • 111-2384
    Spectacle bag, black
    1 * 3 ST - 6.60 SFr.
  •  
    Descrizione
  • 111-2385
    Spectacle pouch
    1 * 1 ST - 11.60 SFr.