VWR®, Dropping Bottles
Bottle only, 30 ml
Codice catalogo 215-3820
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
Soda-lime glass, amber.
- Good chemical and physical properties
- Pipette kit not included
Amber to protect light-sensitive substances, with DIN 18 thread.
Documents
Specifiche
- Capacità:30 ml
- Descrizione:Bottle only
- Seq:05