FOLIC ACID CASEI MEDIUM 1 * 100 g
Catalog # HIMEM543-100G
Anbieter: HIMEDIA
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Anbieter: VWR Chemicals
Acetone ≥99%, TECHNICAL
Anbieter: VWR Chemicals
2-Propanolo ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
2-Propanolo ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Anbieter: VWR Chemicals
Glicerina ≥99.5%, AnalaR NORMAPUR® ACS per analisi, bidistillata
Anbieter: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
2-Propanolo ≥99.5% (by GC), EMPLURA®, Supelco®
Anbieter: Merck
Etanolo assoluto ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
Anbieter: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® per HPLC
Anbieter: VWR Chemicals
Etanolo, GPR RECTAPUR®
Anbieter: VWR Chemicals