Spill kit, spill box
SPILL BOX, chemical, filled box with 10 pads, 9 L
Codice catalogo BRDY813910
Nome del fornitore: Brady
Vai a
Product Details & Documents
These Spill box pads in a convenient dispenser box allows to create multiple spill stations.
- Reusable plastic box can be refilled once all the sorbents have been used
- The bright yellow colour stands out for quick identification and use
The Spill Box is light enough to be hanged to laboratory and storage area walls or to be attached to a forklift. Various application areas: in liquid storage areas, behind the seat of a truck, lab environments, in workstations that deal with liquids.
Documents
BRDY813908_TDS_EN
Scheda dati
Specifiche
- Capacità:9 L
- Descrizione:SPILL BOX, chemical, filled box with 10 pads
- Seq:03