Lens cleaning tissues
Lens cleaning tissue, 200×300 mm
Codice catalogo BINZ146104
Nome del fornitore: Ahlstrom
Vai a
Product Details & Documents
These very fine lens cleaning tissues are made of pure manila vegetable fibre.
- Non abrasive
Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.
12 g/m²
Specifiche
- Descrizione:Lens cleaning tissue
- L×L:200×300 mm
- Seq:06
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



