Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Lens cleaning tissues

Lens cleaning tissues

Lens cleaning tissue, 135×170 mm
Codice catalogo BINZ146102
Nome del fornitore:   Ahlstrom
Lens cleaning tissues
Codice catalogo BINZ146102
Nome del fornitore:  Ahlstrom
Lens cleaning tissues
Lens cleaning tissues

Lens cleaning tissue, 135×170 mm

Price
SFr. 33.50
Log in per disponibilità
1 * 500 ST
Product Details & Documents

These very fine lens cleaning tissues are made of pure manila vegetable fibre.

  • Non abrasive

Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.

12 g/m²

Specifiche
  • Descrizione:
    Lens cleaning tissue
  • L×L:
    135×170 mm
  • Seq:
    04
  • Cat. No.:
    BINZ146102
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.