Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Lens cleaning tissues

Lens cleaning tissues

Lens cleaning tissue, 95×135 mm
Codice catalogo BINZ146100
Nome del fornitore:   Ahlstrom
Lens cleaning tissues
Codice catalogo BINZ146100
Nome del fornitore:  Ahlstrom
Lens cleaning tissues
Lens cleaning tissues

Lens cleaning tissue, 95×135 mm

Price
SFr. 2.80
Log in per disponibilità
1 * 25 ST
Product Details & Documents

These very fine lens cleaning tissues are made of pure manila vegetable fibre.

  • Non abrasive

Lint-free paper for cleaning of optical surfaces such as lenses and mirrors of microscopes, etc.

12 g/m²

Specifiche
  • Descrizione:
    Lens cleaning tissue
  • L×L:
    95×135 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.