Orders LinkContactUsLinkComponent
Sonde
External replacement probe for PT 1000, Ø×L: 5×50 mm, TPX 220-3, −200...+250 °C, T: ±2,0 °C (–200…–100 °C); ±1,0 °C (–100 …–20 °C); ±0,2 °C (–20…+60 °C); ±0,5 °C (+60…+250 °C)
  620-2823
 :   EBRO
Sonde
  620-2823
 :  EBRO
Sonde
Sonde
External replacement probe for PT 1000, Ø×L: 5×50 mm, TPX 220-3, −200...+250 °C, T: ±2,0 °C (–200…–100 °C); ±1,0 °C (–100 …–20 °C); ±0,2 °C (–20…+60 °C); ±0,5 °C (+60…+250 °C)
SFr. 280.00
1 * 1 ST
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
  • Range di misura:
    −200...+250 °C
  • Descrizione:
    External replacement probe for PT 1000, Ø×L: 5×50 mm, TPX 220-3
  • Seq:
    12
  • Accuratezza:
    T: ±2,0 °C (–200…–100 °C); ±1,0 °C (–100 …–20 °C); ±0,2 °C (–20…+60 °C); ±0,5 °C (+60…+250 °C)