Post-mortem scalpel handles
PM8, Blade no's PM60 and PM60B, 185 mm
Nome del fornitore: SWANN MORTON
Vai a
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
- PM40 range with traditional screw fitting
- PM60 range with standard blade fitting
PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Packaging: The PM8 and PM40 stainless steel handles and all blades are supplied individually wrapped.
Specifiche
- Per:Blade no's PM60 and PM60B
- Tipo:PM8
- Lunghezza:185 mm
- Seq:01