Biopsy processing/embedding cassettes, Unisette™ M406
grigio
Codice catalogo SIMPM406-9T
Nome del fornitore: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Codice catalogo SIMPM406-9T
Nome del fornitore: Simport Scientific
Vai a
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Documents
SIMPM406-7_TDS_EN
Scheda dati
Specifiche
- Colore:grigio
- Seq:18