Biopsy processing/embedding cassettes, Unisette™ M406
panna
Codice catalogo SIMPM406-8
Nome del fornitore: Simport Scientific
Biopsy processing/embedding cassettes, Unisette™ M406
Codice catalogo SIMPM406-8
Nome del fornitore: Simport Scientific
Vai a
Product Details & Documents
These disposable biopsy cassettes are designed to hold biopsy specimens during the embedding process. They are also designed as a storage file.
- Moulded from a special high density polymer
- Resistant to the chemical action of most histological solvents
Specifiche
- Colore:panna
- Seq:07