Orders LinkContactUsLinkComponent
Grinding balls for XRD-mill McCrone
Grinding balls for XRD-mill McCrone
Grinding elements set, with 50 grinding elements, Agata, XRD-Mill McCrone
Catalog # RETS02.368.0123
Supplier:   Retsch GmbH
Grinding balls for XRD-mill McCrone
Catalog # RETS02.368.0123
Supplier:  Retsch GmbH
Grinding balls for XRD-mill McCrone
Grinding elements set, with 50 grinding elements, Agata, XRD-Mill McCrone
Price
SFr. 3’080.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Descrizione:
    Grinding elements set, with 50 grinding elements
  • Materiale:
    Agata
  • Seq:
    04
  • Per uso con:
    XRD-Mill McCrone