Bag cutters
Bag cutter
Codice catalogo MICRBC-01
Nome del fornitore: MICRONOVA
Vai a
Product Details & Documents
Plastic cutters for opening raw material packages.
- Automatically retractable safety blade guard
- No blade changes required - eliminates all loose blades
- Ergonomic handle
- Available gamma irradiated
Documents
Specifiche
- Descrizione:Bag cutter
- Seq:01