Permeability cups
Permeability cup with a surface area of 25 cm²
Codice catalogo SIMEVF2201
Nome del fornitore: Industrial Physics
Vai a
Product Details & Documents
Permeability cups are designed to determine the water vapour transmission of paints, varnishes, coatings and related products. Permeability cup is suitable for testing both self-supporting coating and non-self-supporting coatings.
- Anodised aluminium for corrosion resistance and colouring
- Seal ring to prevent turning when closing the cover
- Any deformation to the cup may cause deviation in the result
The wet cup method is the reference for determining water vapour transmission.
Certifications: ISO 7783 (supersedes NF T30-018), ASTM D1653, ASTM E95.
Informazioni per l'ordine: Includes a cup, a seal ring and cover ring.
Documents
TQC Company Video
Video
Specifiche
- Descrizione:Permeability cup with a surface area of 25 cm²
- Seq:02