Orders LinkContactUsLinkComponent
Salmeterol Related Compounds, Reference Standard
Salmeterol Related Compounds, Reference Standard
Salmeterol System Suitability Related Compound H, USP reference standard, Vial, 15 mg
Codice catalogo 1609590.
Nome del fornitore:   USP
Salmeterol Related Compounds, Reference Standard
Codice catalogo 1609590.
Nome del fornitore:  USP
Numero CAS:  
Salmeterol Related Compounds, Reference Standard
Salmeterol System Suitability Related Compound H, USP reference standard, Vial, 15 mg
Numero CAS:  
Price
SFr. 1’560.00
Log in per disponibilità
1 * 15 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Salmeterol System Suitability Related Compound H: Mixture containing salmeterol xinafoate, salmeterol-N-alkyl and salmeterol
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
  • Tipo di conf.:
    Vial
  • Conf:
    15 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Salmeterol System Suitability Related Compound H