Orders LinkContactUsLinkComponent
Insulin, Reference Standards
Insulin, Reference Standards
Insulin Glargine for Peak Identication, USP reference standard, Ampule, 3,2 mg
Codice catalogo 1342060.
Nome del fornitore:   USP
Insulin, Reference Standards
Codice catalogo 1342060.
Nome del fornitore:  USP
Numero CAS:  
Insulin, Reference Standards
Insulin Glargine for Peak Identication, USP reference standard, Ampule, 3,2 mg
Numero CAS:  
Price
1 560.00 SFr.
Log in per disponibilità
1 * 3,2 mg
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Insulin Glargine for Peak Identication: Mixture of Insulin Glargine and 0A-Arg-Insulin Glargine
Certifications: US Pharmacopea Reference Standard
Attenzione: Cold Shipment required
Documents
Specifiche
Specifiche
  • Tipo di conf.:
    Ampule
  • Conf:
    3,2 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Insulin Glargine for Peak Identication
Specifiche