Mobile storage cabinets
Mobile storage cabinet, with 2 drawer of 300 mm height, lockable, 2 drawers, 900×516×740 mm
Codice catalogo KOTT315-00135
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, with 2 drawer of 300 mm height, lockable
- Seq:29
- L×P×A:900×516×740 mm
- Porte:2 drawers