Mobile storage cabinets
Mobile storage cabinet, with 3 drawer of 150 mm height, lockable, 3 drawers, 900×516×590 mm
Codice catalogo KOTT315-00130
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, with 3 drawer of 150 mm height, lockable
- Seq:25
- L×P×A:900×516×590 mm
- Porte:3 drawers