Orders LinkContactUsLinkComponent
Mobile storage cabinets
Mobile storage cabinet, with 4 drawer of 150 mm height, lockable, 4 drawers, 450×516×740 mm
Catalog # KOTT315-00127
Supplier:   KOETTERMANN
Mobile storage cabinets
Catalog # KOTT315-00127
Supplier:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, with 4 drawer of 150 mm height, lockable, 4 drawers, 450×516×740 mm
Price
1 310.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Specifications
  • Descrizione:
    Mobile storage cabinet, with 4 drawer of 150 mm height, lockable
  • Seq:
    19
  • L×P×A:
    450×516×740 mm
  • Porte:
    4 drawers