Mobile storage cabinets
Mobile storage cabinet, lockable, 1 door right, 450×516×740 mm
Codice catalogo KOTT315-00123
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, lockable
- Seq:15
- L×P×A:450×516×740 mm
- Porte:1 door right