Mobile storage cabinets
Mobile storage cabinet with 1 shelf, With front open, 450×500×590 mm
Codice catalogo KOTT315-00107
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet with 1 shelf
- Seq:01
- L×P×A:450×500×590 mm
- Porte:With front open