Orders LinkContactUsLinkComponent
Mobile storage cabinets | Avantor
Mobile storage cabinets
Mobile storage cabinet, with 2 drawer of 150 mm height, 2 doors, 1200×516×740 mm
Katalognr. KOTT315-00104
Anbieter:   KOETTERMANN
Mobile storage cabinets
Katalognr. KOTT315-00104
Anbieter:  KOETTERMANN
Mobile storage cabinets
Mobile storage cabinets
Mobile storage cabinet, with 2 drawer of 150 mm height, 2 doors, 1200×516×740 mm
Preis
1 380.00 SFr.
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Epoxy powder coated steel cabinets
  • Optional locks
  • Fitted with castors
  • Removable shelves
Technische Daten
  • Descrizione:
    Mobile storage cabinet, with 2 drawer of 150 mm height
  • Seq:
    49
  • L×P×A:
    1200×516×740 mm
  • Porte:
    2 doors