Mobile storage cabinets
Mobile storage cabinet, lockable, 2 doors, 1200×516×590 mm
Codice catalogo KOTT315-00096
Nome del fornitore: KOETTERMANN
Vai a
Product Details & Documents
Epoxy powder coated steel cabinets
- Optional locks
- Fitted with castors
- Removable shelves
Specifiche
- Descrizione:Mobile storage cabinet, lockable
- Seq:41
- L×P×A:1200×516×590 mm
- Porte:2 doors