Litio cloruro 99+% ACS
Bottiglia di plastica, 2,5 kg
- :7447-41-8
- :LiCl
- :MFCD00011078
- :42.39 g/mol
- :13,05550
- :1382 °C (760 mmHg)
- :605 °C
- :2,06 g/cm³
- :Ambiente
- :231-212-3
- Appearance:White Crystalline powder or crystals
- Titration Argentometric:≥99 %
- Loss on drying:≤1.0 % (1 g, 105°C, 2 hrs)
- Sulfate (SO4):≤100 ppm
- Barium (Ba):≤30 ppm
- Calcium (Ca):≤100 ppm
- Iron (Fe):≤10 ppm
- Potassium (K):≤100 ppm
- Sodium (Na):≤2000 ppm
- Heavy metals (ICP-OES):≤20 ppm
- Titrable base:≤0.008 meq/g
- Insoluble matter:≤0.01 %
- Tipo di conf.:Bottiglia di plastica
- Conf:2,5 kg



